RIE183967922 Rieke Metals, Inc.

4-Chloro-3-Fluorobenzylzinc chloride

  • MDL number MFCD11226471
  • Molecular Weight : 244.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  
Availability details for
Quantity
  • Availability
  • Add to Cart