RIE183967924 Rieke Metals, Inc.

3,5-Dimethyl-4-methoxyphenethylmagnesium bromide

  • MDL number MFCD09965867
  • Molecular Weight : 267.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/3231

Safety Information

MSDS http://riekemetals.com/products-services/catnum/3231
Availability details for
Quantity
  • Availability
  • Add to Cart