RIE183967926 Rieke Metals, Inc.

4-[4-(2-Methoxyethoxy)phenyl]-4-oxobutyric acid

  • MDL number MFCD09965869
  • Molecular Weight : 252.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7340B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7340B
Availability details for
Quantity
  • Availability
  • Add to Cart