RIE183967927 Rieke Metals, Inc.

5-[4-(2-Methoxyethoxy)phenyl]-5-oxovaleric acid

  • MDL number MFCD09965870
  • Molecular Weight : 266.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7340C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7340C
Availability details for
Quantity
  • Availability
  • Add to Cart