RIE183967928 Rieke Metals, Inc.

6-[4-(2-Methoxyethoxy)phenyl]-6-oxohexanoic acid

  • MDL number MFCD09965871
  • Molecular Weight : 280.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7340D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7340D
Availability details for
Quantity
  • Availability
  • Add to Cart