RIE183967929 Rieke Metals, Inc.

ethyl 4-[4-(2-Methoxyethoxy)phenyl]-4-oxobutyrate

  • MDL number MFCD09965872
  • Molecular Weight : 280.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7840B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7840B
Availability details for
Quantity
  • Availability
  • Add to Cart