RIE183967930 Rieke Metals, Inc.

ethyl 5-[4-(2-Methoxyethoxy)phenyl]-5-oxovalerate

  • MDL number MFCD09965873
  • Molecular Weight : 294.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7840C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7840C
Availability details for
Quantity
  • Availability
  • Add to Cart