RIE183967931 Rieke Metals, Inc.

ethyl 6-[4-(2-Methoxyethoxy)phenyl]-6-oxohexanoate

  • MDL number MFCD09965874
  • Molecular Weight : 308.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7840D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7840D
Availability details for
Quantity
  • Availability
  • Add to Cart