RIE213109870 Rieke Metals, Inc.

3-Pyridylzinc bromide

  • MDL number MFCD11915899
  • Cas Number : 220565-63-9
  • Molecular Weight : 223.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/2282

Safety Information

MSDS http://riekemetals.com/products-services/catnum/2282
Availability details for
Quantity
  • Availability
  • Add to Cart