RIE213109871 Rieke Metals, Inc.

5-Pyrimidylzinc bromide

  • MDL number MFCD11915900
  • Cas Number : 1227941-02-7
  • Molecular Weight : 224.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/2283

Safety Information

MSDS http://riekemetals.com/products-services/catnum/2283
Availability details for
Quantity
  • Availability
  • Add to Cart