RIE213109872 Rieke Metals, Inc.

4-Isoquinolinzinc bromide

  • MDL number MFCD11915901
  • Cas Number : 1207760-37-9
  • Molecular Weight : 273.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/2284
Availability details for
Quantity
  • Availability
  • Add to Cart