RIE213109874 Rieke Metals, Inc.

4-Methyl-3-pyridylzinc bromide

  • MDL number MFCD11915903
  • Cas Number : 1227833-81-9
  • Molecular Weight : 237.40

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/2286

Safety Information

MSDS http://riekemetals.com/products-services/catnum/2286
Availability details for
Quantity
  • Availability
  • Add to Cart