RIE213109875 Rieke Metals, Inc.

2-Methyl-5-pyridylzinc bromide

  • MDL number MFCD11915904
  • Cas Number : 1227833-82-0
  • Molecular Weight : 237.40

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/2287

Safety Information

MSDS http://riekemetals.com/products-services/catnum/2287
Availability details for
Quantity
  • Availability
  • Add to Cart