RIE213109877 Rieke Metals, Inc.

3,4-(Methylenedioxy)phenylzinc iodide

  • MDL number MFCD11915906
  • Cas Number : 1357013-37-6
  • Molecular Weight : 313.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/2289

Safety Information

MSDS http://riekemetals.com/products-services/catnum/2289
Availability details for
Quantity
  • Availability
  • Add to Cart