RIE213109879 Rieke Metals, Inc.

2-Chloro-5-pyridylzinc bromide

  • MDL number MFCD11915908
  • Cas Number : 1227833-84-2
  • Molecular Weight : 257.81

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/2291

Safety Information

MSDS http://riekemetals.com/products-services/catnum/2291
Availability details for
Quantity
  • Availability
  • Add to Cart