RIE240869825 Rieke Metals, Inc.

2-(4-Bromo-2-fluorophenyl)-6-methoxypyridine

  • MDL number MFCD13152459
  • Molecular Weight : 282.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/11075

Safety Information

MSDS http://riekemetals.com/products-services/catnum/11075
Availability details for
Quantity
  • Availability
  • Add to Cart