RIE240869874 Rieke Metals, Inc.

Ethyl 5-(5-methylpyridin-2-yl)thiophene-2-carboxylate

  • MDL number MFCD13152501
  • Molecular Weight : 247.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/11128

Safety Information

MSDS http://riekemetals.com/products-services/catnum/11128
Availability details for
Quantity
  • Availability
  • Add to Cart