RIE240869900 Rieke Metals, Inc.

5,4'-Dimethyl-2,3'-bipyridine

  • MDL number MFCD13152534
  • Molecular Weight : 184.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/11167

Safety Information

MSDS http://riekemetals.com/products-services/catnum/11167
Availability details for
Quantity
  • Availability
  • Add to Cart