RIE240869911 Rieke Metals, Inc.

4-(5-Methyl-2-Pyridyl)aniline

  • MDL number MFCD13152546
  • Molecular Weight : 184.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/11503

Safety Information

MSDS http://riekemetals.com/products-services/catnum/11503
Availability details for
Quantity
  • Availability
  • Add to Cart