RIE240869918 Rieke Metals, Inc.

4-Bromo-2-fluorobenzylzinc bromide

  • MDL number MFCD13152553
  • Molecular Weight : 333.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/2298

Safety Information

MSDS http://riekemetals.com/products-services/catnum/2298
Availability details for
Quantity
  • Availability
  • Add to Cart