RIE240869920 Rieke Metals, Inc.

2,4,5-Trifluorobenzylzinc bromide

  • MDL number MFCD13152555
  • Molecular Weight : 290.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  
Availability details for
Quantity
  • Availability
  • Add to Cart