RIE240869924 Rieke Metals, Inc.

3-Methoxy-4'-trifluoromethylbenzophenone

  • MDL number MFCD13152562
  • Molecular Weight : 280.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3-methoxyphenyl)[4-(trifluoromethyl)phenyl]methanone
Inchi Code 1S/C15H11F3O2/c1-20-13-4-2-3-11(9-13)14(19)10-5-7-12(8-6-10)15(16,17)18/h2-9H,1H3
Inchi Key VUEGCVXHZIMKLZ-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5050-45

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5050-45
Availability details for
Quantity
  • Availability
  • Add to Cart