RIE240869934 Rieke Metals, Inc.

6-Methyl-2-(4-trifluoromethylbenzoyl)pyridine

  • MDL number MFCD13152575
  • Molecular Weight : 265.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (6-methyl-2-pyridinyl)[4-(trifluoromethyl)phenyl]methanone
Inchi Code 1S/C14H10F3NO/c1-9-3-2-4-12(18-9)13(19)10-5-7-11(8-6-10)14(15,16)17/h2-8H,1H3
Inchi Key LIIRVJIZCHBPIJ-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5612-10

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5612-10
Availability details for
Quantity
  • Availability
  • Add to Cart