RIE240869937 Rieke Metals, Inc.

2-(2-Chlorobenzoyl)-6-methylpyridine

  • MDL number MFCD13152579
  • Molecular Weight : 231.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2-chlorophenyl)(6-methyl-2-pyridinyl)methanone
Inchi Code 1S/C13H10ClNO/c1-9-5-4-8-12(15-9)13(16)10-6-2-3-7-11(10)14/h2-8H,1H3
Inchi Key JBVULCNHLABZMP-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5612-14

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5612-14
Availability details for
Quantity
  • Availability
  • Add to Cart