RIE240869941 Rieke Metals, Inc.

2-(3-Bromobenzoyl)-6-methylpyridine

  • MDL number MFCD13152583
  • Molecular Weight : 276.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3-bromophenyl)(6-methyl-2-pyridinyl)methanone
Inchi Code 1S/C13H10BrNO/c1-9-4-2-7-12(15-9)13(16)10-5-3-6-11(14)8-10/h2-8H,1H3
Inchi Key KJYOTHTVVDAHIR-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5612-18

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5612-18
Availability details for
Quantity
  • Availability
  • Add to Cart