RIE240869947 Rieke Metals, Inc.

2-(4-Cyanobenzoyl)-6-methylpyridine

  • MDL number MFCD13152589
  • Molecular Weight : 222.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-[(6-methyl-2-pyridinyl)carbonyl]benzonitrile
Inchi Code 1S/C14H10N2O/c1-10-3-2-4-13(16-10)14(17)12-7-5-11(9-15)6-8-12/h2-8H,1H3
Inchi Key KSRPGDSAKYTEQC-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5612-25

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5612-25
Availability details for
Quantity
  • Availability
  • Add to Cart