RIE240869948 Rieke Metals, Inc.

6-Methyl-2-(3-phenoxybenzoyl)pyridine

  • MDL number MFCD13152590
  • Molecular Weight : 289.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5612-27

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5612-27
Availability details for
Quantity
  • Availability
  • Add to Cart