RIE240869952 Rieke Metals, Inc.

2-(4-Isopropylbenzoyl)-6-methylpyridine

  • MDL number MFCD13152594
  • Molecular Weight : 239.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4-isopropylphenyl)(6-methyl-2-pyridinyl)methanone
Inchi Code 1S/C16H17NO/c1-11(2)13-7-9-14(10-8-13)16(18)15-6-4-5-12(3)17-15/h4-11H,1-3H3
Inchi Key OAQZQBFVAOZOSU-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5612-32

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5612-32
Availability details for
Quantity
  • Availability
  • Add to Cart