RIE240869958 Rieke Metals, Inc.

6-Methyl-2-(4-pentylbenzoyl)-pyridine

  • MDL number MFCD13152600
  • Molecular Weight : 267.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (6-methyl-2-pyridinyl)(4-pentylphenyl)methanone
Inchi Code 1S/C18H21NO/c1-3-4-5-8-15-10-12-16(13-11-15)18(20)17-9-6-7-14(2)19-17/h6-7,9-13H,3-5,8H2,1-2H3
Inchi Key IJEYXUVVKRHJMH-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5612-35d

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5612-35d
Availability details for
Quantity
  • Availability
  • Add to Cart