RIE240869972 Rieke Metals, Inc.

2-(3,5-Difluorobenzoyl)-6-methylpyridine

  • MDL number MFCD13152614
  • Molecular Weight : 233.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5612-42

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5612-42
Availability details for
Quantity
  • Availability
  • Add to Cart