RIE240869978 Rieke Metals, Inc.

2-(3,5-Dichlorobenzoyl)-6-methylpyridine

  • MDL number MFCD13152620
  • Molecular Weight : 266.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5612-48

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5612-48
Availability details for
Quantity
  • Availability
  • Add to Cart