RIE240869988 Rieke Metals, Inc.

2-(2,6-Dimethylbenzoyl)-6-methylpyridine

  • MDL number MFCD13152630
  • Molecular Weight : 225.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2,6-dimethylphenyl)(6-methyl-2-pyridinyl)methanone
Inchi Code 1S/C15H15NO/c1-10-6-4-7-11(2)14(10)15(17)13-9-5-8-12(3)16-13/h4-9H,1-3H3
Inchi Key GXSDSWLYCOPAGM-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5612-58

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5612-58
Availability details for
Quantity
  • Availability
  • Add to Cart