RIE240869989 Rieke Metals, Inc.

2-(3,4-Dimethylbenzoyl)-6-methylpyridine

  • MDL number MFCD13152631
  • Molecular Weight : 225.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5612-59

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5612-59
Availability details for
Quantity
  • Availability
  • Add to Cart