RIE240869992 Rieke Metals, Inc.

6-Methyl-2-(pentafluorobenzoyl)pyridine

  • MDL number MFCD13152634
  • Molecular Weight : 287.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5612-62

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5612-62
Availability details for
Quantity
  • Availability
  • Add to Cart