RIE240869996 Rieke Metals, Inc.

2-(2-Methylbenzoyl)-5-methylpyridine

  • MDL number MFCD13152639
  • Molecular Weight : 211.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2-methylphenyl)(5-methyl-2-pyridinyl)methanone
Inchi Code 1S/C14H13NO/c1-10-7-8-13(15-9-10)14(16)12-6-4-3-5-11(12)2/h3-9H,1-2H3
Inchi Key TUWQEGKNSBUINQ-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5613-05

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5613-05
Availability details for
Quantity
  • Availability
  • Add to Cart