RIE240870000 Rieke Metals, Inc.

5-Methyl-2-(3-trifluoromethylbenzoyl)pyridine

  • MDL number MFCD13152643
  • Molecular Weight : 265.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5613-09

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5613-09
Availability details for
Quantity
  • Availability
  • Add to Cart