RIE240870005 Rieke Metals, Inc.

2-(3-Chlorobenzoyl)-5-methylpyridine

  • MDL number MFCD13152649
  • Molecular Weight : 231.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5613-15

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5613-15
Availability details for
Quantity
  • Availability
  • Add to Cart