RIE240870015 Rieke Metals, Inc.

5-Methyl-2-(3-phenoxybenzoyl)pyridine

  • MDL number MFCD13152658
  • Molecular Weight : 289.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (5-methyl-2-pyridinyl)(3-phenoxyphenyl)methanone
Inchi Code 1S/C19H15NO2/c1-14-10-11-18(20-13-14)19(21)15-6-5-9-17(12-15)22-16-7-3-2-4-8-16/h2-13H,1H3
Inchi Key YMYNGSMJHPIGFU-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5613-27

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5613-27
Availability details for
Quantity
  • Availability
  • Add to Cart