RIE240870017 Rieke Metals, Inc.

5-Methyl-2-(3-nitrobenzoyl)pyridine

  • MDL number MFCD13152660
  • Molecular Weight : 242.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5613-30

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5613-30
Availability details for
Quantity
  • Availability
  • Add to Cart