RIE240870020 Rieke Metals, Inc.

2-(4-Isopropoxybenzoyl)-5-methylpyridine

  • MDL number MFCD13152663
  • Molecular Weight : 255.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5613-33

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5613-33
Availability details for
Quantity
  • Availability
  • Add to Cart