RIE240870024 Rieke Metals, Inc.

2-(4-Butylbenzoyl)-5-methylpyridine

  • MDL number MFCD13152667
  • Molecular Weight : 253.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5613-35C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5613-35C
Availability details for
Quantity
  • Availability
  • Add to Cart