RIE240870028 Rieke Metals, Inc.

2-(4-Ethoxybenzoyl)-5-methylpyridine

  • MDL number MFCD13152671
  • Molecular Weight : 241.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4-ethoxyphenyl)(5-methyl-2-pyridinyl)methanone
Inchi Code 1S/C15H15NO2/c1-3-18-13-7-5-12(6-8-13)15(17)14-9-4-11(2)10-16-14/h4-10H,3H2,1-2H3
Inchi Key MOKFSQLVPLGDKC-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5613-36a

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5613-36a
Availability details for
Quantity
  • Availability
  • Add to Cart