RIE240870030 Rieke Metals, Inc.

2-(4-Butoxybenzoyl)-5-methylpyridine

  • MDL number MFCD13152673
  • Molecular Weight : 269.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5613-36C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5613-36C
Availability details for
Quantity
  • Availability
  • Add to Cart