RIE240870044 Rieke Metals, Inc.

2-(2,3-Dimethoxybenzoyl)-5-methylpyridine

  • MDL number MFCD13152687
  • Molecular Weight : 257.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5613-49

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5613-49
Availability details for
Quantity
  • Availability
  • Add to Cart