RIE240870052 Rieke Metals, Inc.

2-(2,5-Dimethylbenzoyl)-5-methylpyridine

  • MDL number MFCD13152695
  • Molecular Weight : 225.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2,5-dimethylphenyl)(5-methyl-2-pyridinyl)methanone
Inchi Code 1S/C15H15NO/c1-10-4-6-12(3)13(8-10)15(17)14-7-5-11(2)9-16-14/h4-9H,1-3H3
Inchi Key RDMFOKVPLBJYOE-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5613-57

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5613-57
Availability details for
Quantity
  • Availability
  • Add to Cart