RIE240870064 Rieke Metals, Inc.

4-Methyl-2-(2-trifluoromethylbenzoyl)pyridine

  • MDL number MFCD13152707
  • Molecular Weight : 265.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4-methyl-2-pyridinyl)[2-(trifluoromethyl)phenyl]methanone
Inchi Code 1S/C14H10F3NO/c1-9-6-7-18-12(8-9)13(19)10-4-2-3-5-11(10)14(15,16)17/h2-8H,1H3
Inchi Key AOFQDCCEBTVAMO-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5614-08

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5614-08
Availability details for
Quantity
  • Availability
  • Add to Cart