RIE240870066 Rieke Metals, Inc.

4-Methyl-2-(4-trifluoromethylbenzoyl)pyridine

  • MDL number MFCD13152709
  • Molecular Weight : 265.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5614-10

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5614-10
Availability details for
Quantity
  • Availability
  • Add to Cart