RIE240870072 Rieke Metals, Inc.

2-(2-Bromobenzoyl)-4-methylpyridine

  • MDL number MFCD13152716
  • Molecular Weight : 276.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5614-17

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5614-17
Availability details for
Quantity
  • Availability
  • Add to Cart