RIE240870075 Rieke Metals, Inc.

2-(2-Iodobenzoyl)-4-methylpyridine

  • MDL number MFCD13152719
  • Molecular Weight : 323.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5614-20

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5614-20
Availability details for
Quantity
  • Availability
  • Add to Cart