RIE240870078 Rieke Metals, Inc.

2-(3-Cyanobenzoyl)-4-methylpyridine

  • MDL number MFCD13152722
  • Molecular Weight : 222.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-[(4-methyl-2-pyridinyl)carbonyl]benzonitrile
Inchi Code 1S/C14H10N2O/c1-10-5-6-16-13(7-10)14(17)12-4-2-3-11(8-12)9-15/h2-8H,1H3
Inchi Key GVJSPIOUNLISNO-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5614-24

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5614-24
Availability details for
Quantity
  • Availability
  • Add to Cart